missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ Tb Emission Filters, 15 mm
LanthaScreen™ Tb Emission Filters, 15 mm
Brand: Invitrogen™ PV00315
Additional Details : Weight : 0.03000kg
Description
A set of two emission filters (diameter = 15 mm) for use with the LanthaScreen™ Tb assays and products: 495 nm filter with a 10 nm bandpass, and a 520 nm filter with a 25 nm bandwidth.For use primarily with the Perkin Elmer EnVision™ plate reader. Perkin Elmer EnVision™ Empty Filter Holders are required to mount the filters (Perkin Elmer cat# 2100-8110, 10 empty barcoded filter holders for custom filters plus locking rings).
For assistance in setting up your instrument, please contact our Technical Support team in North America at +1 760 603 7200, x40266.
Specifications
Emission Filter | |
LanthaScreen™ | |
15 mm | |
Lanthanide Chelates |
Microplate Reader | |
Set of two filters, shipped and stored at room temperature | |
495/520 nm | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.