Learn More
LACTB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532043
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HCT116. Predicted reactivity: Mouse (82%), Rat (81%), Pig (86%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Endoribonuclease; cleaves preferentially 3' to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3' -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Polyclonal | |
Unconjugated | |
Lactb2 | |
Beta-lactamase-like protein 2; beta-lactamase-like protein 2; endoribonuclease LACTB2; beta-lactamase-like protein 2-like; CGI-83; E430032H21Rik; EGK_19028; endoribonuclease LACTB2; lactamase beta 2; lactamase, beta 2; Lactb2; testicular secretory protein Li 23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q53H82 | |
Lactb2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 288 of Human LACTB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |