missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5100598
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total LACTB.
Endoribonuclease; cleaves preferentially 3' to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3' -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Polyclonal | |
Unconjugated | |
Lactb2 | |
Beta-lactamase-like protein 2; beta-lactamase-like protein 2; endoribonuclease LACTB2; beta-lactamase-like protein 2-like; CGI-83; E430032H21Rik; EGK_19028; endoribonuclease LACTB2; lactamase beta 2; lactamase, beta 2; Lactb2; testicular secretory protein Li 23 | |
Rabbit | |
Affinity chromatography | |
RUO | |
212442, 297768, 51110 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q53H82, Q561R9, Q99KR3 | |
Lactb2 | |
A synthesized peptide derived from human LACTB2(Accession Q53H82), corresponding to amino acid residues D126-D176. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |