missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Monoclonal Antibody (OTI2F9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525465
Additional Details : Weight : 0.01000kg
Description
Endoribonuclease; cleaves preferentially 3' to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3' -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Monoclonal | |
1.1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53H82 | |
Lactb2 | |
Human recombinant protein fragment corresponding to amino acids 78-288 of LACTB2 produced in HEK293T cell | |
100μL | |
Primary | |
Canine, Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F9 | |
Unconjugated | |
Lactb2 | |
Beta-lactamase-like protein 2; beta-lactamase-like protein 2; endoribonuclease LACTB2; beta-lactamase-like protein 2-like; CGI-83; E430032H21Rik; EGK_19028; endoribonuclease LACTB2; lactamase beta 2; lactamase, beta 2; Lactb2; testicular secretory protein Li 23 | |
Mouse | |
Affinity Chromatography | |
RUO | |
486990, 51110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |