Learn More
Thermo Scientific™ Labtainer™ BioProcess Container 2D System, ASI 28 Film, 3 Edge Ports, MPC Insert/Body with C-Flex Tubing/Pinch Clamps
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
898.85 EUR - 1092.37 EUR
Specifications
Port Detail MPC insert and body connectors and injection site
Tubing Style C-Flex, EVA
Sterilization Method Irradiated
Material Ethylene Vinyl Acetate (EVA)
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15969906 View Documents Thermo Scientific™
SS00020-I
5 L 1092.37 EUR
25 units View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-08-2019
Add to basket
15979906 View Documents Thermo Scientific™
SS00021-I
10 L 948.40 EUR
25 units View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-08-2019
Add to basket
15989906 View Documents Thermo Scientific™
SS00022-I
20 L 995.28 EUR
25 units View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-08-2019
Add to basket
15999906 View Documents Thermo Scientific™
SS00023-I
50 L 898.85 EUR
20 units View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-08-2019
Add to basket
Description
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Specifications
MPC insert and body connectors and injection site
C-Flex, EVA
Irradiated
Ethylene Vinyl Acetate (EVA)