Learn More
Amarell™ Laboratory Total Immersion Precision Thermometer
Brand: Amarell™ L26610
Additional Details : Weight : 0.05000kg
Specifications
Red Spirit | |
Total Immersion | |
Glass | |
150°C | |
-10°C to +150°C | |
1°C |
Government testing | |
305 mm | |
Precision Thermometer | |
-10°C | |
Yes |