Learn More
Epredia™ Lab Vision™ Laminin Ab-1 Rabbit Polyclonal Antibody with BSA and azide
Recommended for Immunohistochemistry (Formalin/paraffin). Epredia™ Laminin Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia™ RB-082-RQ
Additional Details : Weight : 0.02000kg
Description
Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Laminin isolated from a rat yolk sac tumor cell line
Positive Control: Skin, Lung, Kidney or Tonsil
Cellular Localization: Basement membrane Ab-1 labels basement membrane surrounding normal vessels, glands, muscles, nerves, fat and deciduas cells separating these from abutting connective tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Laminin Ab-1 | |
Polyclonal | |
Rabbit | |
12 mL | |
Extracellular Matrix | |
Ready-to-Use |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Laminin isolated from a rat yolk sac tumor cell line | |
IVD | |
Primary |