missing translation for 'onlineSavingsMsg'
Learn More

Ku (p70) Ab-4 Mouse Monoclonal Antibody, Epredia™

Recommended for multiple molecular biology techniques, Epredia™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-329-PABX

Additional Details : Weight : 0.00180kg

 View more versions of this product

Product Code. 12691027

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Host Species: Mouse

Clone: N3H10

Isotype: IgG2b

Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.

Epitope: aa 506-541

Immunogen: Human placental extract designated as PSE1-PL

Molecular Weight: 70kDa

Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ku (p70) Ab-4
Monoclonal
1mg/mL
Mouse
200 μL
DNA Repair
Human, Non-human Primate, Xenopus species
IgG2b
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
N3H10
Unconjugated
Human placental extract designated as PSE1-PL
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers