Learn More
Ku (p70) Ab-4 Mouse Monoclonal Antibody, Epredia™
Recommended for multiple molecular biology techniques, Epredia™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-329-R7
Additional Details : Weight : 0.00700kg
Description
The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.Host Species: Mouse
Clone: N3H10
Isotype: IgG2b
Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.
Epitope: aa 506-541
Immunogen: Human placental extract designated as PSE1-PL
Molecular Weight: 70kDa
Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Ku (p70) Ab-4 | |
Monoclonal | |
Unconjugated | |
Human placental extract designated as PSE1-PL | |
RUO | |
Primary | |
Prediluted |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
N3H10 | |
Mouse | |
7 mL | |
DNA Repair | |
Human, Non-human Primate, Xenopus species | |
IgG2b |