missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible resuts in immunohistochemistry applications with Epredia™ CD79a/mb-1 (B-Cell Marker) Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1171-P0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12619206

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.

Host Species: Mouse

Clone: JCB117

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein

Molecular Weight: 50kDa

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD79a/mb-1 (B-Cell Marker) Ab-2
Monoclonal
200μg/mL
Mouse
100 μL
B-Cell
Human
IgG2a κ
Immunohistochemistry (Paraffin)
JCB117
Unconjugated
Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers