Learn More
Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible resuts in immunohistochemistry applications with Epredia™ CD79a/mb-1 (B-Cell Marker) Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1171-P
Additional Details : Weight : 0.00250kg
Description
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.Host Species: Mouse
Clone: JCB117
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Extracellular domain
Immunogen: Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein
Molecular Weight: 50kDa
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD79a/mb-1 (B-Cell Marker) Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
B-Cell | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
JCB117 | |
Unconjugated | |
Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein | |
IVD | |
Primary | |
Concentrated |