Learn More
Epredia™ Lab Vision™ CD163 Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD163 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1103-S
Additional Details : Weight : 0.00250kg
Description
CD163 antigen is a type I membrane protein also known as M130 antigen, Ber-Mac3, Ki-M8 or SM4. CD163 is restricted in its expression to the monocytic/macrophage lineage. It is present on all circulating monocytes and most tissue macrophages except those found in the mantle zone and germinal centers of lymphoid follicles, interdigitating reticulum cells and Langerhan's cells.Host Species: Mouse
Clone: 10D6
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Domains 1 to 4 of the N-terminal region
Immunogen: Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
CD163 Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding of domains 1-4 of the N-terminal region of human CD163 | |
1 mL | |
Hematopoietic | |
Human | |
IgG1 |
Immunohistochemistry (Paraffin) | |
10D6 | |
Mouse | |
Placenta | |
RUO | |
Primary | |
Concentrated |