Learn More
CD155/PVR (Poliovirus Receptor) Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunofluorescence and flow cytometry applications.
Brand: Epredia MS-465-P0
Additional Details : Weight : 1.00000kg
Description
PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from a single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or α and κ,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbones, respectively. Two other splice variants, Β and γ, encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues.Host Species: Mouse
Clone: D171
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with pig, dog, rabbit, rat, hamster and mouse. Others not known.
Epitope: aa 35-50
Immunogen: HeLa cells
Molecular Weight: 60-80kDa
Positive Control: HeLa, SV 80, HEp-2, RD, A431, HL-60, U-937 cells; Monkey Vero, GMK, or TC-7 cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunofluorescence
- Flow Cytometry
- Inhibits Replication of Poliovirus in HeLa Cells
Note:
USA: RUO; Int'l: RUO
Specifications
CD155 Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Cell Adhesion | |
Human, Non-human Primate | |
IgG1 |
Immunofluorescence | |
D171 | |
Unconjugated | |
HeLa cells | |
RUO | |
Primary | |
Concentrated |