Learn More
TexWrite™ 22 Cleanroom Bond Paper
Shop All Texwipe Products
Click to view available options
Color:
Blue
Unit Size:
Pack of 2500
Description
- No organic fillers
- No natural latex binders
- High strength and excellent heat resistance
- Excellent toner adhesion
- Cleanroom packaged
- Precision-cut edges
- Very low particle counts and sodium levels
- Reduced ionic and metal contamination
- Compatible with toner and offset printing
- Autoclavable for use in sterile environment
Specifications
Specifications
| Color | Blue |
| Dimensions (L x W) | 297 x 210 mm |
| Length (Metric) | 297 mm |
| Width (Metric) | 210 mm |
| Autoclavable | Autoclavable |
| For Use With (Application) | Standard-duty and high-speed laser printers and photocopiers |
| Size | A4 |