Learn More
L-β-Homoproline hydrochloride, 98% , Thermo Scientific Chemicals
Catalog Number p-3770753
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
53912-85-9
C6H12ClNO2
165.617
MFCD07363484
VQDACVOAOJQTPR-JEDNCBNOSA-N
s-2-pyrrolidin-2-yl acetic acid hydrochloride, l-beta-homoproline hydrochloride, l-beta-homoproline hcl, 2-2s-pyrrolidin-2-yl acetic acid hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2s, s-2-2-pyrrolidinyl acetic acid hydrochloride, s-3-pyrrolidinecarboxylic acid hydrochloride, 2s-pyrrolidin-2-ylacetic acid hydrochloride, l-beta-homoproline-hcl, s-3-pyrrol-idinecarboxylic acid hydrochloride
2761540
2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride
C1CC(NC1)CC(=O)O.Cl
53912-85-9
C6H12ClNO2
165.617
MFCD07363484
VQDACVOAOJQTPR-JEDNCBNOSA-N
s-2-pyrrolidin-2-yl acetic acid hydrochloride, l-beta-homoproline hydrochloride, l-beta-homoproline hcl, 2-2s-pyrrolidin-2-yl acetic acid hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2s, s-2-2-pyrrolidinyl acetic acid hydrochloride, s-3-pyrrolidinecarboxylic acid hydrochloride, 2s-pyrrolidin-2-ylacetic acid hydrochloride, l-beta-homoproline-hcl, s-3-pyrrol-idinecarboxylic acid hydrochloride
2761540
2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride
C1CC(NC1)CC(=O)O.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
NMR Consistent with structure
Packaging Glass Bottle
Specific Rotation +31° to +35° (20°C, 589nm) (c=1, H2O)
Formula Weight 165.62
Percent Purity ≥97.5%
Quantity 250 mg
Physical Form Crystalline Powder
Chemical Name or Material L-β-Homoproline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L-beta-Homoproline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.