missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine hydrochloride monohydrate, MP Biomedicals™
Click to view available options
Quantity:
100 g
1 kg
5 kg
Specifications
Formula Weight | 175.64 |
Grade | Cell Culture Reagent |
Quantity | 100 g |
Physical Form | Crystalline |
Chemical Name or Material | L-Cysteine Hydrochloride Monohydrate, Precursor in the food, pharmaceutical, personal care industries |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'