missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine hydrochloride hydrate, 98.5-101.0%, Thermo Scientific Chemicals

Catalog Number p-3734466
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
345909-32-2
C3H8ClNO2S
157.61
MFCD00065606
IFQSXNOEEPCSLW-QYLAHFPANA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
Cl.N[C@@H](CS)C(O)=O
This item is not returnable. View return policy
345909-32-2
C3H8ClNO2S
157.61
MFCD00065606
IFQSXNOEEPCSLW-QYLAHFPANA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
Cl.N[C@@H](CS)C(O)=O

L-cysteine hydrochloride hydrate, CA S# 7048-04-6, also known as L-cysteine hydrochloride monohydrate, is a soluble salt of the non-essential amino acid, L-cysteine. It is commonly used as a nucleophile in several chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• L-cysteine hydrochloride hydrate is a non-essential proteinogenic amino acid commonly used in chemical reactions, for instance, the thiol side chain in cysteine often participates in enzymatic reactions as a nucleophile.

Application

• L-cysteine hydrochloride hydrate plays an important role as a histidine ammonia-lyase inhibitor with the ability to interfere with the action of histidine ammonia-lyase.
• In chemical reactions, L-cysteine hydrochloride hydrate is used as a raw material for the production of S-carboxymethyl-L-cysteine, L-cysteine base, and N-acetyl-L-cysteine.
• This compound also has antioxidant activity.
• It has been used in laboratory protocols to separate dorsal root ganglion neurons.
• This compound is applied to study the inhibitory effect of L-cysteine hydrochloride hydrate in preventing enzymatic browning in apples.

TRUSTED_SUSTAINABILITY
Melting Point 175°C
Ammonium 100ppm max.
Color White or Colorless
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 98.5-101.0%
Infrared Spectrum Authentic
Linear Formula HCl·xH2O
Iron (Fe) 20ppm max.
Beilstein 04, 506
Loss on Drying 8.0% to 12.0% (24 hrs, 5mm Hg)
Merck Index 14, 2781
Packaging Glass Bottle
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Specific Rotation +5.5° to +7.0° (20°C, 589nm) (c=8, 6N HCl)
Sulfate 300ppm max.
Formula Weight 157.61
Percent Purity 98.5 to 101.0% (on dried substance)
Sulfated Ash 0.1% max.
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Cysteine hydrochloride hydrate
Show More Show Less
compliance-icons
Product Identifier
  • L-Cysteine hydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'