missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ku (p70) Ab-4 Mouse Monoclonal Antibody, Epredia™
Recommended for multiple molecular biology techniques, Epredia™ Ku (p70) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
149.00 EUR - 974.10 EUR
Specifications
Antigen | Ku (p70) Ab-4 |
---|---|
Clone | N3H10 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12651027
|
Epredia
MS-329-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12681027
|
Epredia
MS-329-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12691027
|
Epredia
MS-329-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12671027
|
Epredia
MS-329-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12661027
|
Epredia
MS-329-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12631037
|
Epredia
MS-329-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.Host Species: Mouse
Clone: N3H10
Isotype: IgG2b
Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.
Epitope: aa 506-541
Immunogen: Human placental extract designated as PSE1-PL
Molecular Weight: 70kDa
Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Ku (p70) Ab-4 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Human, Non-human Primate, Xenopus species | |
IgG2b |
N3H10 | |
Monoclonal | |
Mouse | |
DNA Repair | |
Human placental extract designated as PSE1-PL | |
Primary |