Learn More
DWK Life Sciences Kimble™ Kontes™ Schlenk Vacuum Tube with HI-VAC Valve
Suitable for use with K216050 or K216060 Manifolds
277.00 EUR - 366.00 EUR
Specifications
Product Type | Vacuum Tube |
---|---|
Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
Material | Borosilicate Glass |
For Use With | 216050 or 216060 manifolds |
Includes | Hi-Vac™ valve, valve plug |
Product Code | Brand | Height (English) | Overall Length (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Overall Length (English) | Width (English) | Price | Quantity & Availability | |||
11815115
|
DWK Life Sciences
218710-0015 |
3.93 in. | 6.49 in. | 2.55 in. |
277.00 EUR
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
11848532
|
DWK Life Sciences
218710-0100 |
5.9 in. | 5.9 in. | 3.14 in. |
366.00 EUR
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
11880014
|
DWK Life Sciences
218710-0025 |
3.93 in. | 6.49 in. | 2.75 in. |
277.00 EUR
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
11838532
|
DWK Life Sciences
218710-0050 |
4.72 in. | 4.72 in. | 2.95 in. |
323.00 EUR
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
Description
- Medium wall borosilicate glass
Specifications
Vacuum Tube | |
Borosilicate Glass | |
Hi-Vac™ valve, valve plug |
USP Type I, ASTM E438, Type I, Class A | |
216050 or 216060 manifolds |