Learn More
DWK Life Sciences Kimble™ Kontes™ Airless-Ware Glass Plug Gas Inlet Adapter
Inlet Adapter for Vacuum and AIRLESS-WARE™, with a choice of 14/20 or 24/40 standard taper joints.
Brand: DWK Life Sciences 211300-1420
Additional Details : Weight : 0.50000kg
Description
- Inner Standard Taper joints
- 211300 series has a glass plug
- Manufactured from 33 expansion, low extractable borosilicate glass
- Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
USP Type I, ASTM E438, Type I, Class A | |
98mm | |
Borosilicate Glass | |
14/20 Inner | |
60mm |
3.85 in. | |
Patented 809000 pressure/vacuum retainer | |
Inlet Adapter | |
2.36 in. |