Learn More
Mettler Toledo™ Knick Adapter for Sensors
For use with Knick portable meters.
Brand: Mettler Toledo™ 30247853
Additional Details : Weight : 0.10000kg
Description
- For sensors with 12mm shaft diameter
- Provided individually
Specifications
Knick adapter | |
For sensors with 12mm shaft diameter | |
Knick portable meters |