Learn More
DWK Life Sciences Kimble™ ShellVial with N-51A Borosilicate Glass Plain Top
Available with or without closure
75.40 EUR
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
Style | Plain top |
Quantity | 144 Pk. |
Product Code | Brand | Volume (English) | Height (English) | Closure Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Height (English) | Closure Type | Price | Quantity & Availability | |||
11853183
|
DWK Life Sciences
60965D-1 |
1 dr. | 1.77 in. | With Titeseal™ Open-bottom Plug Style Plastic Closures, Unattached |
75.40 EUR
Pack of 144 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||
Description
- TITESEAL™ vials
- Plain tops
- Item 60965D1 comes with open-bottom plug style plastic closures, unattached
- Manufactured from 51 expansion borosilicate glass
- Conforms to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Clear | |
Borosilicate Glass | |
Plain top | |
144 Pk. |