Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
Brand: DWK Life Sciences 270504000
Additional Details : Weight : 1.00000kg
Description
- Heavy walled
- Necks tooled for uniform stopper fit with a thick flat flange
- Without side tabulation
- Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
ASTM, E1406, Type I | |
4 L | |
150 to 500 mL | |
14.96 in. | |
20.8 cm |
8.18 in. | |
Borosilicate Glass | |
250 mL | |
38 cm | |
12 |