missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ HPLC Economy 3-Hole Ultra-Ware Cap Systems

With plastic coating that blocks virtually all UV light up to 385 nm, preventing photodegradation of light sensitive mobile phases

Brand:  DWK Life Sciences 9539302002

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11762095

  • 748.00 EUR / Each


Estimated Shipment
04-08-2021
Add to basket

Description

Description

  • Made of type I, Class A borosilicate glass with a plastic safety coating
  • Cap body is made of PTFE
  • Supplied with TFE/propylene o-ring and polypropylene screw collar
  • Connecting threads on top of cap use standard 1/4-28 flangeless fittings
  • Two ports have 1/8 in. through holes, third port has 1/16 in. through hole

Specifications

Specifications

Bottle
Economy Three Hole Cap System with 2 Liter Reservoir
PTFE
2 L
TFE/Propylene o-ring and Polypropylene screw collar, 2L reservoir
1 Cs.
Videos
SDS
Documents