Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE End Fitting for Chromaflex Columns
Click to view available options
Diameter (Metric) Inner:
10 mm
250 mm
Unit Size:
Each
Description
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
- PTFE Ending Fitting with Bed Support
- 1.0cm, 2.5cm and 4.8cm options
- Requires attachment with CTFE adapter 420804-0001, not included
Specifications
Specifications
| Type | End Fitting |
| Diameter (Metric) Inner | 250 mm |
| Includes | Bed supports |
| For Use With | Chromaflex™ column |