Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
With or without fritted disc
Brand:  DWK Life Sciences 420320-0256
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11837423
  • 264.00 EUR / Each
Estimated Shipment: 11-11-2024
Description
  • Borosilicate glass
  • Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
  • Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
Specifications
41 mm
Adsorption/Ion Exchange Chromatography Column
790 mL
Borosilicate Glass
Many separations in adsorption and ion exchange chromatography
40 to 60 μm
600 mm
Fritted disc