Learn More
DWK Life Sciences Kimble™ KIMAX™ Colorware™ Flasks
Colorful enamel graduations enhance readability and indicate approximate volumes
Brand:  DWK Life Sciences 26500R-250
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11812330
  • 113.00 EUR / Pack of 12
Estimated Shipment: 03-01-2025
Description
Colorful enamel graduations enhance readability and indicate approximate volumes
  • Narrow mouth
  • Durable, matte-finish marking spot provides good writing surface
  • Without stoppers
Compliance
Designed from ASTM™ E 1404, Type I, Class 1 requirements
Specifications
Specifications
Rubber
3.2 in.
Borosilicate Glass
25 mL
13 cm
Narrow
6
Raging Red Ceramic Enamel scale graduations
250 mL
50 to 225 mL
5.11 in.
Matte Finish Marking
82 mm