Learn More
Kimberly-Clark Professional™ Kimtech™ Pure G3 Sterile Latex Gloves
269.96 EUR valid until 2024-05-31
Use promo code "21679" to get your promotional price.
388.47 EUR valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Use promo code "21679" to get your promotional price.
388.47 EUR valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
10
Description
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Recommended for Class 10/ISO 4 or higher cleanrooms.
- Fully sterilized latex material provides high levels of contamination protection
- Hand-specific, disposable and a natural color
- Textured palm and fingertips enhance grip and tactile sensitivity
- Beaded cuffs add strength to the gloves reducing the risk of tearing
- Easy donning and doffing
- Made from latex sterilized with gamma radiation and containing no powder
- Suitable for use in Class 10/ISO 4 sterile cleanrooms
Specifications
Certifications/Compliance | ASTM D3577, ASTM D5151, ASTM D412, ASTM D573, ASTM D3767, ASTM D6319, IEST-RP-CC005, ASTM D5712 |
Certifications/Compliance (Europe) | EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016 |
Cleanroom Class | Class 10 (ISO 4) |
AQL | 1.5 |
Color | Natural |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Hand | Specific |
Material | Latex |
Powdered | Powder Free |