Learn More
DWK Life Sciences KECK™ Quick Pressure Coupling KK, complete with tubing parts
For safe connection of 2 hoses.
Brand:  DWK Life Sciences 286340304
106.40 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.16200kg
 View more versions of this product
Product Code. 11726978
  • 112.00 EUR / Pack of 10
Estimated Shipment: 16-05-2024
Description
The KECK™ quick pressure coupling with hose parts quickly, safely and tightly connects 2 hoses and is easy to connect or disconnect with just 1/4 turn. Available in 2 sizes: 2 x 4.5 mm and 2 x 9 mm.
  • Complete with hose part
  • Quick pressure coupling with two hose parts
  • Easy connection and disconnection with only 1/4 turn
Specifications
Specifications
10
Tight Connection of Two Hose Ends
Tight connection of two hose ends
9 mm