Learn More
DWK Life Sciences KECK™ Adapter KA, complete, from glass thread to hose connection plastic
KECK™ adapters protect glass threads from damage.
Brand:  DWK Life Sciences 286345302
67.74 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11860560
  • 71.30 EUR / Pack of 10
Estimated Shipment: 07-06-2024
Description
KECK™ adapters for glass threads are available for sizes GL 14 and RD 14. They protect the thread from damage and ensure a fast, secure and tight connection. The threaded cap is applied directly to the external thread of the connection piece. The hose part can be quickly connected to the threaded cap with a 1/4 turn and loosened again.
  • Screw the threaded cap directly onto the external thread of the connection piece
  • Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
  • They protect the thread from damage and ensure a fast, secure and tight connection.
  • Available for glass threads sizes GL 14 and RD 14
Specifications
Specifications
10
Connection of a Hose Piece with a Threaded Cap
Connection of a hose piece with a threaded cap
RD 14, diameter 9.0 mm