Learn More
Potassium Chloride Solution, SI Analytics™
Catalog Number 11994749
Click to view available options
Quantity:
1000 mL
SI Analytics™ Potassium Chloride Solution is used in the application and preparation of electrolyte solutions.
  • Mixture, consisting of the following components: Water, potassium chloride
  • Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
  • Must not be disposed together with household garbage. Do not allow product to reach sewage system
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
  • Wash off residuals with water
Melting Point −12.8°C
Boiling Point 107.2°C
Color Colorless
Density 1.15g/cm3
Concentration 3 M
pH 7
Quantity 1000 mL
Solution Type Electrolyte
Odor Odorless
Packaging DURAN™ Bottle
Sterility Sterile
Solubility Information Fully miscible in water