Learn More
Kartell™ K70 Silicon Hoses
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
Brand:  Kartell™ 0392600
Additional Details : Weight : 0.73000kg
 View more versions of this product
Product Code. 15732855
  • 85.80 EUR / 10m
Estimated Shipment: 24-05-2024
Description
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
  • Flexible and transparent
  • FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved
  • Can withstand temperatures of -60°C to 200°C
Specifications
Specifications
FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved
Silicone
10 m
10 m
200°C
7 mm
11 mm
-60°C
Transparent