Learn More
Cole-Parmer™ K Type Extension Cables
Use these extension cables for a variety of applications. Cole-Parmer™ K Type Extension Cables feature 20-gauge (0.032 in. dia) Thermocouple Wire with PVC/PVC Insulation.
Brand: Cole-Parmer™ 08516-30
Additional Details : Weight : 0.12000kg
Description
- Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow and type T-blue
Specifications
ANSI | |
Extension Cable | |
3 m |