Learn More
Jenway™ Interference Filters
Jenway™ Interference Filters are fluorometer filters suitable for narrower wavelength requirements.
Brand: Jenway™ 10089-74
Additional Details : Weight : 0.10000kg
Description
- Fluorometer filter
- Normally used as the emission filter where a much narrower range of wavelength is required.
- Allow a specified wavelength of light ±5nm through the sample.
Specifications
Interference Filter | |
Pore Size: 300 nanometer | |
300 nm |