missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Jackson Safety™ G80 Chemical Resistant Neoprene Gloves

Yellow and blue, hand-specific gloves providing reliable protection against chemical and basic mechanical hazards.

Brand:  Kimberly-Clark™ Professional 38744

Additional Details : Weight : 1.09000kg

 View more versions of this product

Product Code. 15396748

  • 186.66 EUR / 120 boxes


Estimated Shipment
31-01-2018
Add to basket

Description

Description

  • Suitable for various applications and offer reliable protection against wide range of chemicals
  • Ideal for: providing PPE Category 3 protection, basic mechanical protection in many different segments when handling chemicals, oils, grease, acids, caustics or solvents
  • Hand specific gloves of 0.70mm thickness
  • Made of neoprene over latex formulation to maximize protection
  • Enhances flexibility with straight cuff and honeycomb structure on the palm to provide good grip in wet and dry conditions
  • Cotton flocked liner for easy donning and extra comfort in extended use
  • Chlorinated for low extractable proteins and residual chemicals
Specifications

Specifications

42.4 x 16.8cm
10
0.70mm
Yellow and Blue
Neoprene
5.04kg
Gloves
42.4cm
Videos