Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
Brand: GPE Scientific™ JC35
Additional Details : Weight : 0.05000kg
Description
- Spring clip design
- Securely fasten J Young spherical joints
Specifications
Spring Clips for J Young Spherical Joints | |
35/20 spherical joints |