Learn More
Isopropyl bromoacetate, 97%, Thermo Scientific Chemicals
Catalog Number p-151582
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
29921-57-1
C5H9BrO2
181.03
MFCD00009886
JCWLEWKPXYZHGQ-UHFFFAOYSA-N
isopropyl bromoacetate, acetic acid, bromo-, 1-methylethyl ester, acetic acid, 2-bromo-, 1-methylethyl ester, isopropyl 2-bromoacetate, brch2c o och ch3 2, bromoacetic acid isopropyl ester, i-propyl bromoacetate, acmc-1ciu8, isopropyl .alpha.-bromoacetate
121593
propan-2-yl 2-bromoacetate
CC(C)OC(=O)CBr
29921-57-1
C5H9BrO2
181.03
MFCD00009886
JCWLEWKPXYZHGQ-UHFFFAOYSA-N
isopropyl bromoacetate, acetic acid, bromo-, 1-methylethyl ester, acetic acid, 2-bromo-, 1-methylethyl ester, isopropyl 2-bromoacetate, brch2c o och ch3 2, bromoacetic acid isopropyl ester, i-propyl bromoacetate, acmc-1ciu8, isopropyl .alpha.-bromoacetate
121593
propan-2-yl 2-bromoacetate
CC(C)OC(=O)CBr
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.3990g/mL
Boiling Point 59.0°C to 61.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CO2CH(CH3)2
Refractive Index 1.4440 to 1.4460
Quantity 25 g
Specific Gravity 1.399
Formula Weight 181.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isopropyl bromoacetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Isopropyl bromoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.