Learn More
Grant Instruments™ IQ/OQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Brand:  Grant Instruments™ IQOQ TC120
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15142615
  • 705.00 EUR / Each
Estimated Shipment: 30-08-2024
Description
  • Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
  • Operational Qualification (OQ): instrument is functioning in accordance with its specification
  • Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
Specifications
Specifications
Grant TC120 Stirred Thermostatic Bath/Circulator
IQ/OQ Documentation