missing translation for 'onlineSavingsMsg'
Learn More

Iodine R4 solution EU Pharmacopoeia, Fisher Chemical™

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Brand:  Fisher Chemical RP 106 1000

Additional Details : Weight : 1.00000kg


Product Code. 12940444

  • 164.00 EUR / 1000mL
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Glass Bottle
EU Ph Ref: 1045806
Iodine solution R4.
1000 mL
Eu. Ph.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iodine solution R4. European Pharmacopoeia Ref: 1045806

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P314-Get medical advice/attention if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : potassium iodide, hydrochloric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers