Learn More
Invitrogen™ Vivid Colors™ pcDNA™6.2/C-EmGFP-DEST Vector
Description
Includes
Each vector is supplied as supercoiled and lyophilized plasmid DNA.
- These vectors contain the next-generation EGFP, Emerald Green Fluorescent Protein (EmGFP) for simple, non-invasive detection of recombinant protein
- Fluorescent proteins have been humanized for optimal mammalian expression
- Vectors offer the CMV promoter for high-level expression of the recombinant fluorescent fusion protein
- Fuse to the C-terminus of the protein
Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Cloning, Constitutive Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifications
Specifications
| Constitutive or Inducible System | Constitutive |
| Delivery Type | Transfection |
| Promoter | CMV |
| Product Type | Mammalian Expression Vector |
| Reporter Gene | GFP (EmGFP) |
| Selection Agent (Eukaryotic) | Blasticidin |
| Content And Storage | Each vector is supplied as supercoiled and lyophilized plasmid DNA. Store at -20°C after resuspended. Guaranteed stable for 6 months when properly stored. |
| Protein Tag | GFP (EmGFP) |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Show More |