Learn More
Invitrogen™ pcDNA™6.2/V5-PL-DEST Mammalian Expression Vector
Designed for high-level, constitutive expression in variety of mammalian cell lines
Brand: Invitrogen™ 12537162
339.24 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.03000kg
Includes: All destination vectors are provided lyophilized and supercoiled.
Requires : Gateway™ entry clone, appropriate Gateway™ LR Clonase enzyme mix, and reaction buffer.
Description
- Promoterless version of pcDNA™6.2/V5-DEST vector
- attR sites for Gateway cloning
- Compatible with MultiSite Gateway Pro kits
- C-terminal V5 tag for easy detection
- Blasticidin resistance gene for efficient stable selection
- Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli
- Gateway Cloning Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an UltimateORF Clone
Cloning,Constitutive Expression,Gateway Cloning,Mammalian Expression,Multi-Gene Expression,Multisite Gateway,Protein Expression,Proteins, Expression, Isolation and Analysis
Specifications
Transfection | |
None (Promoterless) | |
All destination vectors are provided lyophilized and supercoiled. | |
C-terminal | |
Gateway | |
Gateway™, pcDNA™ |
MultiSite Gateway™ Pro kits (e.g. cat. no. 12537-100) | |
Mammalian Expression Vector | |
Mammalian | |
V5 Epitope Tag | |
pcDNA, pDEST, MultiSite Gateway Vectors | |
Clone your own Promoter, Multi-Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.