Learn More
Memmert™ Interior Socket
Interior IP68 socket (for temperature restriction +70°C option) 230V, 2. A. Cannot be switched individually, moisture tight IP68
Brand: Memmert™ IPP750ecoR3
Additional Details : Weight : 0.01100kg
Description
- Accessory
- Ampacity 230 V/2.2 A
- Can be switched off with the On/Off switch
Specifications
IPP750eco | |
230 V |