Learn More
Eppendorf™ Innova™ 40 Incubator Shaker
Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 40 Incubator Shakers provide very broad temperature capabilities for culturing a wide variety of organisms in flasks up to 3 liters.
Brand:  Eppendorf™ M1299-0082
Additional Details : Weight : 63.00000kg
 View more versions of this product
Product Code. 12508886
  • 11585.00 EUR / Each
Estimated Shipment: 08-08-2024
Description
  • Exceptionally quiet operation provides a favorable work environment
  • Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years
  • Compact benchtop model – fits easily on the bench and holds flasks up to 3L
  • Clear cover allows easy viewing of the cultures and lifts easily to access all samples
  • Versatile accessory platform (sold separately) accommodates flasks up to 3L as well as test tube racks and Microplate holders
  • Certified. Meets cETL, UL, CSA and CE standards
Specifications
Specifications
19 mm
230 V, 50 Hz
3 L
50 Hz
101.6 cm
Innova 40
25 to 500 rpm
230 V
Incubator Shaker
76.2 cm
55.9 cm
0.01 to 99:59 hr.