Learn More
Aqualytic™ Incubator, thermostatic control +2°C to +40°C, with LED display, 175L capacity
Brand:  Aqualytic™ 438220
Additional Details : Weight : 51.00000kg
Product Code. 15207928
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Temperature range +2°C to +40°C, adjustable in steps of 0.1°C
  • Low power consumption
  • Illuminated LED Display
  • Power sockets inside incubator
  • Delivered with bottom grid
No. racks: 3No. Sockets: 5Volume: 175L