Learn More
Fisherbrand™ Immersion Basket with Handle
Faciliates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11776188
  • 181.00 EUR / Each
Estimated Shipment
06-04-2021
Add to basket
Description
Description
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
9.4 L Elmasonic S 80 (H) Ultrasonic Baths
76.2 mm
Stainless Steel
Basket
454.66 mm
106.68 mm