Learn More
Duran™ Imhoff Type Sedimentation Cone
Determine the volume of settleable solids from a known volume of waste. Duran™ Imhoff Type Sedimentation Cone can be used for fresh or saline water, or domestic or industrial wastewater.
Brand: Duran™ 214015403
Additional Details : Weight : 0.62900kg
Description
- Graduated
- Durable Duran™ construction
- Determine the volume of settleable solids from a known volume of waste
- Can be used with fresh or saline water, or domestic or industrial wastewater
- Can be used to measure the volume of sand in a sample of well water