Learn More
Duran™ Imhoff Type Sedimentation Cone
Determine the volume of settleable solids from a known volume of waste. Duran™ Imhoff Type Sedimentation Cone can be used for fresh or saline water, or domestic or industrial wastewater.
Brand:  Duran™ 214015403
Additional Details : Weight : 0.62900kg
Product Code. 11975048
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Graduated
  • Durable Duran™ construction
  • Determine the volume of settleable solids from a known volume of waste
  • Can be used with fresh or saline water, or domestic or industrial wastewater
  • Can be used to measure the volume of sand in a sample of well water