Learn More
Rat IgM Isotype Control (eBRM), APC, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 17-4341-82
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control APC has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgM Control antibody specifically detects Rat IgM in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgM | |
eBRM | |
APC | |
Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rat | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgM κ |
For Research Use Only.