Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 46-4732-82
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: Mouse IgG2b Isotype Control PerCP-eFluor® 710 has been reported for use in flow cytometric analysis. Applications Tested: Mouse IgG2b K Isotype Control PerCP-eFluor® 710 has been tested by flow ctometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG2b kappa | |
eBMG2b | |
PerCP-eFluor 710 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.