Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), PE, eBioscience™, Invitrogen™
Mouse Isotype Control
Brand: Affymetrix eBioscience 12-4732-42
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG2b isotype control has been reported for use in flow cytometric analysis. Applications Tested: This Mouse IgG2b K Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and can be used at the same concentration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2b kappa | |
eBMG2b | |
PE | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
For Research Use Only.