Learn More
IgG (γ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody, Epredia™
IgG (γ-Heavy Chain) Ab-1, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states ensuring the ideal tests are always available.
Brand: Epredia RB-1432-R7
Additional Details : Weight : 0.01200kg
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgG isolated from a pool of normal human sera
Molecular Weight: 50 kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgG (gamma heavy Chain) Ab-1 | |
Polyclonal | |
Unconjugated | |
IgG isolated from a pool of normal human sera | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Rabbit | |
7 mL | |
Immunoglobulins | |
Human | |
IgG |